Anti-VIM Mouse Monoclonal Antibody [clone: LN-6]
ABNOMAB13451
: Abnova
- Antibody type:Primary
- Antigen name:Vimentin
- Antigen symbol:VIM
- Clonality:Monoclonal
- Clone:LN-6
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Human
- Gene ID:7431
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against native human VIM.
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: LN-6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human