Anti-EPCAM/1120 Mouse Monoclonal Antibody [clone: EGP40/1120]
ABNOMAB13330
: Abnova
- Antibody type:Primary
- Antigen symbol:EPCAM/1120
- Clonality:Monoclonal
- Clone:EGP40/1120
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Epitope:Full-length
- Gene ID:4072
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human EPCAM.
Type: Primary
Antigen: EPCAM/1120
Clonality: Monoclonal
Clone: EGP40/1120
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human