Anti-AR Mouse Monoclonal Antibody [clone: AR441]
Katalog # ABNOMAB13322
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:AR
- Clonality:Monoclonal
- Clone:AR441
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:367
- Sequence:STEDTAEYSPFKGGYTK
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of human AR.
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: AR441
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human