Anti-Epstein-Barr virus EBNA-1 Goat Polyclonal Antibody
Catalog # ABNOPAB29840
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Epstein-Barr virus EBNA-1
- Clonality:Polyclonal
- Host:Goat
- Reactivity:EBV (Epstein-Barr virus)
- Size:500 μL
- Pk:500 µl
Specifications
About this item
Goat polyclonal antibody raised against Epstein-Barr virus (EBV) EBNA-1.
Type: Primary
Antigen: Epstein-Barr virus EBNA-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Epstein-Barr virus (EBV)