Anti-Hepatitis B surface antigen Goat Polyclonal Antibody
ABNOPAB29849
: Abnova
- Antibody type:Primary
- Antigen name:Hepatitis B surface antigen
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Hepatitis B virus (HBV)
- Size:1 mg
- Pk:1 mg
Goat polyclonal antibody raised against hepatitis B surface antigen (HBsAg).
Type: Primary
Antigen: Hepatitis B surface antigen
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Hepatitis B virus (HBV)