Anti-gC-2 Mouse Monoclonal Antibody [clone: P1124]
ABNOMAB13585
: Abnova
- Antibody type:Primary
- Antigen name:Herpes simplex virus type 2 gC-2
- Antigen symbol:gC-2
- Clonality:Monoclonal
- Clone:P1124
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Herpes simplex virus
- Size:500 μg
- Pk:500 µG
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gC-2.
Type: Primary
Antigen: gC-2
Clonality: Monoclonal
Clone: P1124
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Herpes simplex virus (HSV)