Anti-gC2 Mouse Monoclonal Antibody [clone: H1A022]
ABNOMAB13577
: Abnova
- Antibody type:Primary
- Antigen name:Herpes simplex virus type 1 gC2
- Antigen symbol:gC2
- Clonality:Monoclonal
- Clone:H1A022
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Herpes simplex virus
- Environmentally Preferable:
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against herpes simplex virus type 1 (HSV-1) gC.
Type: Primary
Antigen: gC2
Clonality: Monoclonal
Clone: H1A022
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Herpes simplex virus (HSV)