Order Entry
Denmark
ContactUsLinkComponent
Anti-gD Mouse Monoclonal Antibody [clone: P1103]
Anti-gD Mouse Monoclonal Antibody [clone: P1103]
  ABNOMAB13569
 :  Abnova
Anti-gD Mouse Monoclonal Antibody [clone: P1103]
  ABNOMAB13569
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Herpes simplex virus gD
  • Antigen symbol:
    gD
  • Clonality:
    Monoclonal
  • Clone:
    P1103
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Herpes simplex virus
  • Size:
    500 μg
  • Pk:
    500 µG

 

 

Mouse monoclonal antibody raised against herpes simplex virus (HSV) gD.

Type: Primary
Antigen: gD
Clonality: Monoclonal
Clone: P1103
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Herpes simplex virus (HSV)