Anti-CMV ICP22 Mouse Monoclonal Antibody [clone: CA010]
Catalog # ABNOMAB13519
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytomegalovirus ICP22
- Antigen symbol:CMV ICP22
- Clonality:Monoclonal
- Clone:CA010
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Cytomegalovirus
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against cytomegalovirus (CMV) ICP22.
Type: Primary
Antigen: CMV ICP22
Clonality: Monoclonal
Clone: CA010
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cytomegalovirus (CMV)