Anti-Clostridium difficile toxin B Mouse Monoclonal Antibody [clone: HM020]
ABNOMAB13508
: Abnova
- Antibody type:Primary
- Antigen name:Clostridium difficile toxin B
- Clonality:Monoclonal
- Clone:HM020
- Host:Mouse
- Isotype:IgG1
- Reactivity:Clostridium spp.
- Size:1 mg
- Pk:1 mg
Mouse monoclonal antibody raised against Clostridium difficile toxin B.
Type: Primary
Antigen: Clostridium difficile toxin B
Clonality: Monoclonal
Clone: HM020
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Clostridium Species