Order Entry
Denmark
ContactUsLinkComponent
Anti-Escherichia coli O157 Mouse Monoclonal Antibody [clone: HM016]
Anti-Escherichia coli O157 Mouse Monoclonal Antibody [clone: HM016]
Catalog # ABNOMAB13544
Supplier:  Abnova
Anti-Escherichia coli O157 Mouse Monoclonal Antibody [clone: HM016]
Catalog # ABNOMAB13544
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Escherichia coli O157
  • Clonality:
    Monoclonal
  • Clone:
    HM016
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    E. coli
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Mouse monoclonal antibody raised against Escherichia coli O157.

Type: Primary
Antigen: Escherichia coli O157
Clonality: Monoclonal
Clone: HM016
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Escherichia coli