Anti-Canine distemper virus Mouse Monoclonal Antibody [clone: HM196]
Catalog # ABNOMAB13540
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Canine distemper virus
- Clonality:Monoclonal
- Clone:HM196
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Virus
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against canine distemper virus.
Type: Primary
Antigen: Canine distemper virus
Clonality: Monoclonal
Clone: HM196
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Viruses