Anti-CMV pp65 Mouse Monoclonal Antibody [clone: CA003]
ABNOMAB13529
: Abnova
- Antibody type:Primary
- Antigen name:Cytomegalovirus pp65
- Antigen symbol:CMV pp65
- Clonality:Monoclonal
- Clone:CA003
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Cytomegalovirus
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against cytomegalovirus (CMV) pp65.
Type: Primary
Antigen: CMV pp65
Clonality: Monoclonal
Clone: CA003
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Cytomegalovirus (CMV)