Anti-Rotavirus Mouse Monoclonal Antibody [clone: J340]
ABNOMAB13645
: Abnova
- Antistof type:Primary
- Antigen name:Rotavirus
- Clonality:Monoclonal
- Clone:J340
- Vært:Mouse
- Isotype:IgG2a
- Reactivity:Virus
- Størrelse:1 mg
- Pakn:1 mg
Mouse monoclonal antibody raised against rotavirus.
Type: Primary
Antigen: Rotavirus
Clonality: Monoclonal
Clone: J340
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Viruses