Anti-CHRND Rabbit Polyclonal Antibody
Catalog # ABNOPAB29996
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CHRND
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:1144
- Sequence:LTLGLLAALAVCGSWGLNEEERLIRHLFQEKGYNKELRPVAHKEESVDVA
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of human CHRND.
Type: Primary
Antigen: CHRND
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human