Anti-CDX4 Rabbit Polyclonal Antibody
ABNOPAB29992
: Abnova
- Antibody type:Primary
- Antigen name:Caudal type homeobox 4
- Antigen symbol:CDX4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:1046
- Sequence:TDHQRLELEKEFHCNRYITIQRKSELAVNLGLSERQVKIWFQNRRAKERK
- Size:100 μL
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of human CDX4.
Type: Primary
Antigen: CDX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human