Anti-CYP4A22 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30016
Leverandør: Abnova
Specifikationer
- Antibody type:Primary
- Antigen symbol:CYP4A22
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:284541
- Sequence:MSVSVLSPSRRLGGVSGILQVTSLLILLLLLIKAAQLYLHRQWLLKALQQ
- Size:100 μL
- Pk:100 µl
Specifikationer
Om denne vare
Rabbit polyclonal antibody raised against synthetic peptide of human CYP4A22.
Type: Primary
Antigen: CYP4A22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human