Order Entry
Denmark
ContactUsLinkComponent
 
Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30120
Supplier:  Abnova
undefined
Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30120
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GTF2I repeat domain containing 1
  • Antigen symbol:
    GTF2IRD1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    9569
  • Sequence:
    TFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQV
  • Size:
    100 μL
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of human GTF2IRD1.

Type: Primary
Antigen: GTF2IRD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human