Anti-FOXA3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30081
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:FOXA3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:3171
- Sequence:FENGCYLRRQKRFKLEEKVKKGGSGAATTTRNGTGSAASTTTPAATVTSP
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of human FOXA3.
Type: Primary
Antigen: FOXA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human