Order Entry
Denmark
ContactUsLinkComponent
Hazardous products shippers
Hazardous products shippers
Catalog # CORTFRIGIPACK4L
Supplier:  E3 Cortex
Hazardous products shippers
Catalog # CORTFRIGIPACK4L
Supplier:  E3 Cortex

Specifications

  • Capacity:
    4 l
  • Description:
    Hazardous products shippers Frigipack
  • Pk:
    1

Specifications

About this item

Isothermal packaging allowing for shipment at low temperature of products sensitive to temperature variations.

  • Available in standard version
  • Supplied with preassembled isothermal barrier in 60 mm PSE