Order Entry
Denmark
ContactUsLinkComponent
Anti-HLA-DR Mouse Monoclonal Antibody (APC-Cyanine7) [clone: L243]
Anti-HLA-DR Mouse Monoclonal Antibody (APC-Cyanine7) [clone: L243]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-HLA-DR Mouse Monoclonal Antibody (APC-Cyanine7) [clone: L243]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-9952-T025
  • 25-9952-T100
  • Antigen symbol:
  • HLA-DR
  • HLA-DR
  • Antigen name:
  • MHC Class II antigens, HLA-DR
  • MHC Class II antigens, HLA-DR
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • L243
  • L243
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3122
  • 3122
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together