Work positioning lanyard, Miller® HandZup®
Supplier: HONEYWELL SAFETY
Certificates
About this item
The ergonomic HandZup® is a unique, patented work positioning system (EN 358) that leaves the hands free and enables the operator to work in complete safety.
- Easy to use, ensuring adjustment is quick, accurate and flexible for greater comfort while working
- Miller HandZup® work positioning lanyard available in 2 m, 3 m or 4 m length
- With a twist lock karabiner and SNAP HOOK and protective sheath
Certifications: EN 358
Frequently Bought Together