Mop frame, stainless steel, Clino® CR
PPSA2009061
- Description:Mop frame Clino CR, stainless steel with ClinoConnect Lock
- Length:400 mm
- Seq:02
- Pk:1
Robust and durable through stainless steel mop frame.
- Autoclavable
- Contactless release of the mop with MopDrop
- With quick-change system ClinoConnect