Order Entry
Denmark
ContactUsLinkComponent
Test Kits, chlorine and pH, Comparator 2000+
Test Kits, chlorine and pH, Comparator 2000+
Catalog # 705-0353
Supplier:  Lovibond
CAS Number:  
undefined
Test Kits, chlorine and pH, Comparator 2000+
Catalog # 705-0353
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Chlorine and pH test kit
  • Seq:
    01
  • Type:
    AF 116A
  • Range:
    0,1 - 1,0 mg/l (Cl2);
    6,8 - 8,4 pH
Specifications
About this item
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Delivery: Test kit is supplied in a robust carrying case containing 1× chlorine comparator disc, 1× pH comparator disc, 4× comparator vials, 10 ml, 13 mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.