Glass wool
Supplier: AGILENT HEWLETT PACKARD
Certificates
About this item
Frequently Bought Together
Diethyl ether ≥99.5% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Disposable cuvettes
Supplier: Brand
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
Sulphuric acid 95-97%, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Column cutter
Supplier: PerkinElmer
Disodium UDP-glucose ≥90%, white powder
Supplier: MP Biomedicals
Glass wool
Supplier: AGILENT HEWLETT PACKARD