Order Entry
Denmark
Orders LinkContactUsLinkComponent
Indirect impact tester
Indirect impact tester
Impact tester incl. base plate assembly, guide tube, release collar, two punches (Ø = 12,7 and 15,9 mm)
Katalog # SIMESP1880
Leverandør:   Industrial Physics
Indirect impact tester
Katalog # SIMESP1880
Leverandør:  Industrial Physics
Indirect impact tester
Indirect impact tester
Impact tester incl. base plate assembly, guide tube, release collar, two punches (Ø = 12,7 and 15,9 mm)
Price
17.920,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Impact testers used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Certifications: ISO 6272-2 and ASTM D2794
Documents
Specifikationer
  • Description:
    Impact tester incl. base plate assembly, guide tube, release collar, two punches (Ø = 12,7 and 15,9 mm)
  • Seq:
    05
Anbefalinger