Order Entry
Denmark
ContactUsLinkComponent
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
Supplier:  VWR Chemicals
Certificates
undefined
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Chloride:
    <0.018%
  • Heavy Metals (as Pb):
    ≤3 ppm
  • Nitrogen:
    ≤0.01%
  • pH (30%, Water) @25°C:
    4.5 - 6.5
  • Purity:
    ≥99%
  • Residue on Ignition:
    ≤0.05%
  • Sulphate:
    ≤0.024%
  • Water (KF):
    ≤11%

Specification Test Results

Related Information

Frequently Bought Together