Occupational shoes, clogs, Easy, 37622/37623
Catalog # 114-7262
Supplier: Abeba
Specifications
- Shoe size:38
- Colour:White/blue
- Pk:1 PAIR
Specifications
About this item
ESD clog made from smooth leather with a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
- Replaceable acc Wave insoles; orthopaedic insoles possbile
- Sanitized® treatment reduces odour and bacterial growth
- TPU sole with high slip resistance
Suitable for use in kitchens and in food-processing industries.
Colour: White/blue, white/red
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; OB, FO, E, A, SRC