HPTLC plates silica gel 60, modified CN, DIOL, NH₂, glass backed
Supplier: Merck
Certificates
About this item
CN- and DIOL-modified silica plates are moderately polar that are suitable for both normal and reversed phase systems. The amino modified NH₂ plate provides weak basic ion exchange characteristics with special selectivity for charged compounds offering an alternative to PEI cellulose.