Order Entry
Denmark
ContactUsLinkComponent
GLS screw thread reductions, BOLA
GLS screw thread reductions, BOLA
  BOHLD785-24
 :  Bohlender
undefined
GLS screw thread reductions, BOLA
  BOHLD785-24
 :  Bohlender
  • Description:
    From thread GLS 80 to reducing thread GL 45
  • Pk:
    1
This blue screw cap is made of PP with GLS threads. with movable reduction body made of PTFE for transition to GL threads. The body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable. The product is only exposed to PTFE.
  • Applicable for connecting or inserting tubing, tubes or probes
  • Temperature resistance from −20 to +140 ºC
  • Very good chemical resistance
  • Autoclavable at 121 ºC
  • FDA conformity