Order Entry
Denmark
Orders LinkContactUsLinkComponent
Disposable tip adapters
Disposable tip adapters
Supplier:   HAMILTON BONADUZ
Disposable tip adapters
Supplier:  HAMILTON BONADUZ
Certificates
Disposable tip adapters
Disposable tip adapters
2 Options Available Below
Product Details & Documents
These adapters enable the connection of a standard 1 ml or 200 µl disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used in conjunction the PB600-1 and the disposable tip adapters can be used as a small volume repeating pipette.
  • Ideal for eliminating the possibility for cross contamination
Product Family Options
Product Information
  • Availability
    Price
  • HAMI31330
    Disposable tip adapter, CTFE, 100 µl
    Disposable tip adapter, CTFE, 100 µl
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 341,00 kr
    341,00 kr
    1 * 1 Items
       341,00 kr
  • HAMI31331
    Disposable tip adapter, CTFE, 1 ml
    Disposable tip adapter, CTFE, 1 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 565,00 kr
    565,00 kr
    1 * 1 Items
       565,00 kr