0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Homogenisers
Dispersing Tools
Dispersing tool for WiseTis® homoginisers
Dispersing tool for WiseTis® homoginisers
WITGDH.WHG02108
:
witeg Labortechnik
Dispersing tool for WiseTis® homoginisers
WITGDH.WHG02108
:
witeg Labortechnik
Volume:
1 - 50 ml
Description:
Homogeniser tool, HT1008
Length:
138 mm
Pk:
1
Stator/rotor Ø:
8,0/6,1 mm
For:
HG-15A, WiseTis®