Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beaker, nickel
Beaker, nickel
Certificates
3 Options Available Below
Product Details & Documents
Beaker, Nickel, with pourout lip.
  • Excellent corrosion and temperature resistance
  • Smooth surfaces for fast, easy cleaning
Nickel is completely resistant to phosphoric acid as well as being highly resistant to the corrosive effect of the strongest alkalies. Nickel, however, is less than satisfactory when used for salt solutions containing oxidants such as ferric chloride or solutions of mineral acids containing oxidising salts.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 125 ml
    67 mm
    56 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.480,00 kr
    2.480,00 kr
    1 * 1 Items
       2.480,00 kr
  • 250 ml
    92 mm
    63 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.070,00 kr
    2.070,00 kr
    1 * 1 Items
       2.070,00 kr
  • 2000 ml
    171 mm
    137 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11.470,00 kr
    11.470,00 kr
    1 * 1 Items
       11.470,00 kr