Heat sealers, Eppendorf HeatSealer S100/S200
Supplier: EPPENDORF
Certificates
About this item
The Eppendorf HeatSealer Family offers reliable and effective sealing of a variety of plates. When working with plates, only a tight and reproducible seal gives predictable and reproducible assay performance. Every laboratory works with different kinds of plates and the HeatSealer will work with most common formats — from large deepwell plates to low profile PCR plates. Specific adapter sets are available to cover almost any plate format.
- Hermetic sealing of multiwell plates
- Minimises evaporation in PCR, reducing cross contamination
- Seals 96- and 384-well plates
- Integrated thermostat prevents overheating
- Improved mechanics for easier sealing
Delivery: Includes PCR plate adapter. Other plate adapters are available separately.
Specifications
- Catalog No:
- EPPE5391000.001
- EPPE5392000.005
- Model:
- S100
- S200
- Operating humidity range:
- 20 - 80 % (non condensing)
- 20 - 80 % (non condensing)
- Operating temperature:
- 18...30 °C
- 18...30 °C
- Temperature set range:
- 170 °C
- 125...200 °C (in 1 °C increments)
- Time set point range:
- -
- 1 to 9 s in 0,5 s increments
- W×D×H:
- 220×250×230 mm
- 220×321×425 mm
- Weight:
- 4 kg
- 7,2 kg
Specifications
Frequently Bought Together