Order Entry
Denmark
ContactUsLinkComponent
 
 
4-Bromo-2-methylbenzenesulphonyl chloride 97%
  ACRO451780100
 :  
undefined
 
4-Bromo-2-methylbenzenesulphonyl chloride 97%
  ACRO451780100
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Yellow to brown Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    62 to 66 °C
  • GC:
    ≥96.0 %

 

 

 :
 :269.55
 :164 °C (15 mmHg)
 :62…67 °C
 :Ambient
 :3261
 :8,II
 :GHS05
 :139937-37-4
 :C₇H₆BrClO₂S
 :MFCD00219927