Order Entry
Denmark
ContactUsLinkComponent
Bromelain (juice) (from Stem)
Bromelain (juice) (from Stem)
Certificates
Bromelain (juice) (from Stem)

Specifications

  • CAS No.:
    9001-00-7
  • Source:
    Stem
  • Storage conditions:
    Refrigerator
  • Form:
    Cream to light brown powder
  • MDL No.:
    MFCD00081423
  • Enzyme name:
    Bromelain (juice)

Specifications

Related Information

Frequently Bought Together