



Lead(II) perchlorate trihydrate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-62-8
:
Formula:Cl₂O₈Pb·3H₂O
MDL Number:MFCD00149817
:460.15 g/mol
Melting Pt:100 °C (dec.)
Density:2.6 g/cm³
UN:1470
ADR:5.1,II
:GHS07
:GHS08
:GHS09
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Complexometric:97.0 to 102.0 %
- pH:3.0 to 5.0 (5% in water, 25.0°C)
- Chloride (Cl):≤0.01 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together