
Benzyl alcohol ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Boiling Pt:205 °C (760 mmHg)
Melting Pt:–15 °C
Density:1.044 g/cm³
Flash Pt:96 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- GC:≥99.0 %
- Color scale:≤20 APHA
- Ash:≤0.005 %
Specification Test Results
Frequently Bought Together