
Diammonium hydrogen citrate 98+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3012-65-5
:
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068
:226.19 g/mol
Density:1.5 g/cm³
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Residue after ignition:≤0.01 %
- Titration with NaOH:98.0 to 103.0 %
- Chloride (Cl):≤0.001 %
- Oxalate (C2O4):Passes test (Limit about 0.05%)
- Phosphate (PO4):≤5 ppm
- Iron (Fe):≤0.001 %
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.005 %
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together