Order Entry
Denmark
ContactUsLinkComponent
Crystal violet ACS
Crystal violet ACS
Certificates
undefined
Crystal violet ACS

Specification Test Results

  • Appearance:
    Dark green Powder or crystals
  • Assay:
    ≥90.0 % (on dry substance)
  • Loss on drying:
    ≤7.5 %
  • Absorbance characteristics:
    (0.98-1.9) Passes test
  • Sensitivity:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together