
Diammonium hydrogen citrate 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3012-65-5
:
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068
:226.19 g/mol
Density:1.5 g/cm³
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Solubility:Clear colorless
- UV:(1 M in water)
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤0.001 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together