Potassium thiocyanate 99% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassium rhodanide
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Boiling Pt:500 °C (1013 hPa)
Melting Pt:170…179 °C
Density:1.886 g/cm³
:97.18 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystals
- Titration Argentometric:≥98.5 %
- pH:5.3 to 8.7
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Ammonium (NH4):≤0.003 %
- Copper (Cu):≤0.0005 %
- Iron (Fe):≤0.0005 %
- Lead (Pb):≤0.001 %
- Sodium (Na):≤0.005 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together