
L-Dihydroorotic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5988-19-2
:
Formula:C₅H₆N₂O₄
MDL Number:MFCD00085339
:158.113 g/mol
Melting Pt:254…255 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:253 to 257 °C
- HPLC:≥98.5 %
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together