Urea 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Boiling Pt:197 °C (1013 hPa)
Melting Pt:131…135 °C
Density:1,34 g/cm³ (20 °C)
:60.06 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:132 to 135 °C
- Loss on drying:≤0.1 % (1 g, 105 °C, 1 h)
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.1 % (800 °C)
- UV:(6 M in H2O)
- pH:6.0 to 9.5 (20 °C, 10% in H2O)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤0.05 %
- Sulfate (SO4):≤0.05 %
- Ammonium (NH4):≤0.05 %
Specification Test Results
Frequently Bought Together