Order Entry
Denmark
ContactUsLinkComponent
 
Ethylene glycol, anhydrous 99.8%, AcroSeal®
 
undefined
Ethylene glycol, anhydrous 99.8%, AcroSeal®
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless Viscous liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.75 %

 

Frequently Bought Together