
Tetrabutylammonium chloride xhydrate 96%
:37451-68-6
:88641-55-8
:C₁₆H₃₆ClN·xH₂O
:MFCD00011635
:277.92 g/mol (anhy)
:37…39 °C
:GHS07
:Ambient
- Appearance:Off-white Low melting solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥95.0 % (On dried substance)
- Water:≤10.0 % (K.F.)
Frequently Bought Together