
Cesium methanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2550-61-0
:
Formula:CH₃CsO₃S
MDL Number:MFCD00075523
Melting Pt:262…264 °C
:227.997 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless
Specification Test Results
Frequently Bought Together