Propylene oxide 99.5%, AcroSeal®, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
CAS Number:75-56-9
:
Formula:C₃H₆O
MDL Number:MFCD00005126
Boiling Pt:34 °C
Melting Pt:–112 °C
Density:0.83 g/cm³
Molecular Weight:58.08 g/mol
Flash Pt:–37 °C
Storage Temperature:Ambient
ADR:3 I
Merck Index:12,08041
UN:1280
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3650 to 1.3670 (20°C, 589 nm)
- Water:≤0.02 %
- GC:≥99.4 %
- Color scale:≤10 APHA
- Aldehyde (C3H6O):≤0.01
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO43086
SDS
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together