Cyanogen bromide 3 M in dichloromethane, AcroSeal®
Catalog # 428911000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Strength:2.95 to 3.30 M
- GC:≥96.0 %
Specification Test Results
About this item
:
:105.92 g/mol
Density:1.44 g/cm³
Storage Temperature:Refrigerator
UN:2927
ADR:6.1,I
:GHS06
:GHS08
:GHS05
:GHS09
CAS Number:506-68-3
Formula:CBrN
MDL Number:MFCD00011597