Order Entry
Denmark
ContactUsLinkComponent
Methyl cellulose, 400 cP (2% H₂O)
Methyl cellulose, 400 cP (2% H₂O)
Certificates
undefined
Methyl cellulose, 400 cP (2% H₂O)

Specification Test Results

  • Appearance:
    White to yellowish-white to greyish-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 %
  • Viscosity:
    300 to 560 cP (2 % in water at 20°C)

Specification Test Results

Related Information

Frequently Bought Together